missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) chloride tetrahydrate, MilliporeSigma™
Specifications
Specifications
| Quantity | 250 g |
| Solubility Information | Soluble in water: 1600g/L at 10°C |
| Formula Weight | 198.83 |
| Percent Purity | ≥98% |
| Odor | Odorless |
| Packaging | Plastic Bottle |
| Density | 1.93g/cm3 at 20°C |
| Chemical Name or Material | Iron(II) chloride tetrahydrate |
Safety and Handling
| Hazard Category | H302: Harmful if swallowed. |
| Hazard Statement | H315: Causes skin irritation. |
| Precautionary Statement | H318: Causes serious eye damage. |
| EINECSNumber | 231-843-4 |
| RTECSNumber | NO5600000 |
| Recommended Storage | Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. |