missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide hydrate

Catalog No. p-7049914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3113314 25 g
AA3113322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3113314 Supplier Thermo Scientific Chemicals Supplier No. 03113314
Only null left

CAS: 13463-12-2 | Br2Fe | 215.65 g/mol

It is widely as a pharmaceutical intermediate.It is a useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13463-12-2
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
IUPAC Name λ²-iron(2+) dibromide
SMILES [Fe++].[Br-].[Br-]

Specifications

Melting Point 27°C
Color Green
Physical Form Lump
Quantity 25 g
Linear Formula FeBr2·xH2O
UN Number UN3260
Solubility Information It is soluble in water,soluble in other solvents such as THF,methanol,ethanol.
Formula Weight 215.67 (Anhydrous)
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 3.9 g/mL
Chemical Name or Material Iron(II) bromide hydrate
Show More Show Less
Hazard Category H314-H318
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-168-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only