missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide, anhydrous, 98% min

Catalog No. p-4407470
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3115406 5 g
AA3115414 25 g
AA3115422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3115406 Supplier Thermo Scientific Chemicals Supplier No. 03115406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7789-46-0 | Br2Fe | 215.65 g/mol

Iron(II) bromide is used as an intermediate in the synthesis porphryins. It is also used as weak reducing agents. It is used as precursor for the preparation of [(C2H5)4N]2FeBr4 by reacting with tetraethylammonium bromide. It is also involved in the synthesis of iron pyrite nanocrystals and nanocrystalline thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-46-0
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
IUPAC Name λ²-iron(2+) dibromide
SMILES [Fe++].[Br-].[Br-]

Specifications

Melting Point 684°C
Color Yellow
Physical Form Granules
Quantity 5 g
Assay Percent Range 98% min.
Solubility Information Soluble in water,methanol,ethanol and tetrahydrofuran.
Formula Weight 215.67
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Yellow granules
Density 4.636 g/cm3
Chemical Name or Material Iron(II) bromide, anhydrous
Show More Show Less
Hazard Category H314-H318
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packing Group: III
EINECSNumber 232-168-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only