missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Acetate 90.0+%, TCI America™

Catalog No. p-7131586
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07655G 5 g
I076525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07655G Supplier TCI America Supplier No. I07655G
Only null left

Chemical Identifiers

CAS 3094-87-9
Molecular Formula C4H6FeO4
Molecular Weight (g/mol) 173.93
MDL Number MFCD00058909
InChI Key LNOZJRCUHSPCDZ-UHFFFAOYSA-L
Synonym iron ii acetate, ferrous acetate, iron diacetate, iron di acetate, iron 2+ acetate, acetic acid, iron 2+ salt, acetic acid, iron 2+ salt 2:1, unii-l80i7m6d3q, ferrousacetate
PubChem CID 18344
IUPAC Name λ²-iron(2+) diacetate
SMILES [Fe++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 5 g
Formula Weight 173.93
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iron(II) Acetate
EINECSNumber (2)-0693
RTECSNumber AI3850000
TSCA Yes