missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) Acetate 90.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 173.93 |
| Percent Purity | ≥90.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Iron(II) Acetate |
Safety and Handling
| EINECSNumber | (2)-0693 |
| RTECSNumber | AI3850000 |
| TSCA | Yes |