missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron, For Analysis Reduced, Particle Size 10um EMSURE™, MilliporeSigma™
$132.39 - $443.66
Chemical Identifiers
| CAS | 7439-89-6 |
|---|---|
| Molecular Formula | Fe |
| Molecular Weight (g/mol) | 55.85 |
| MDL Number | MFCD00010999 |
| InChI Key | XEEYBQQBJWHFJM-UHFFFAOYSA-N |
| Synonym | ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m |
| PubChem CID | 23925 |
| ChEBI | CHEBI:82664 |
| IUPAC Name | iron |
| SMILES | [Fe] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1038190100
|
Technidata
1038190100 |
100 g | Plastic bottle |
Each for $132.39
|
|
||||
|
M1038190500
|
Technidata
1038190500 |
500 g | Plastic bottle |
Each for $443.66
|
|
||||
Chemical Identifiers
| 7439-89-6 | |
| 55.85 | |
| XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
| 23925 | |
| iron |
| Fe | |
| MFCD00010999 | |
| ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
| CHEBI:82664 | |
| [Fe] |
Specifications
| 7439-89-6 | |
| 100 g | |
| Plastic bottle | |
| Fe | |
| UN 3089 | |
| XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
| iron | |
| 23925 | |
| 55.85. (molar mass) |
| Gray solid | |
| Odorless | |
| Solid | |
| MFCD00010999 | |
| ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
| [Fe] | |
| 55.85 | |
| CHEBI:82664 | |
| Iron |
Safety and Handling
H228: Flammable solid
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-096-4