missing translation for 'onlineSavingsMsg'
Learn More

Iron, For Analysis Reduced, Particle Size 10um EMSURE™, MilliporeSigma™

Catalog No. M1038190100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1038190100 100 g Plastic bottle
M1038190500 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1038190100 Supplier Technidata Supplier No. 1.03819.0100

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Appearance Gray solid
Quantity 100 g
Odor Odorless
Packaging Plastic bottle
Form Solid
UN Number UN 3089
Formula Weight 55.85. (molar mass)
Chemical Name or Material Iron
Hazard Category H228: Flammable solid
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-096-4