missing translation for 'onlineSavingsMsg'
Learn More

Iron foil, 0.5mm (0.02in) thick, 99.99% (metals basis)

Form: approx. 2.5g/25x25mm | CAS: 7439-89-6 | Fe | 55.85 g/mol

Supplier:  Thermo Scientific Chemicals 040495FI

 View more versions of this product

Catalog No. AA40495FI


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
7439-89-6
7.87g/cm3 at 20°C
99.99% (Metals basis)
Fe
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
55.85
CHEBI:82664
Iron
Gray foil
50 x 50 mm
Foil
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925

Chemical Identifiers

7439-89-6
55.85
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 231-096-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only