missing translation for 'onlineSavingsMsg'
Learn More

Iron foil, 0.025mm (0.001in) thick, hard, 99.5% (metals basis)

Catalog No. AA44687GT
Change view
Click to view available options
Quantity:
150 x 150 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44687GT 150 x 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA44687GT Supplier Thermo Scientific Chemicals Supplier No. 044687GT
Only null left

CAS: 7439-89-6 | Fe | 55.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Quantity 150 x 150 mm
Weight ≈4.4g/150x150mm
Form Foil
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Iron foil, 0.025mm (0.001 in.) thick, Hard
Hazard Category H500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only