missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Iron Filings

Catalog No. S95970
Change view
Click to view available options
Product Type:
Coarse Iron Filing
Fine Iron Filing, Plastic Jar
Fine Iron Filing, Sprinkle Jar
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S95970 Fine Iron Filing, Plastic Jar
S95969 Fine Iron Filing, Sprinkle Jar
S95968 Coarse Iron Filing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S95970 Supplier Eisco™ Supplier No. PH0799C

Iron Filings are great for magnetism demonstrations.

  • Choose from coarse or fine grain size
  • Convenient packaging choices

Specifications

Description Iron Filings
Material 100% Iron
Shape Cylindrical
For Use With (Application) For Magnetism Demonstrations
Grade 9 to 12
Color Grey
Product Type Fine Iron Filing, Plastic Jar
Includes GHS-compliant label printed with safety and storage information
Quantity 250 g
Age 8 years old