missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Iron Filings

Catalog No. p-4416760
Change view
Click to view available options
Quantity:
500 g
Packaging:
Lab Grade - Fine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25369B 500 g Lab Grade - Fine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25369B Supplier Fisher Science Education™ Supplier No. S25369B

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Black to Gray
Density 7.86g/mL at 20°C
Quantity 500 g
Packaging Lab Grade - Fine
Assay Percent Range >97% w/v
Linear Formula Fe
Formula Weight 55.85
Chemical Name or Material Iron, Filings
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 4.1, II, : Flammable Solid