missing translation for 'onlineSavingsMsg'
Learn More

Iron arsenide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog No. AA3619009
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3619009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3619009 Supplier Thermo Scientific Chemicals Supplier No. 03619009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12005-88-8 | Molecular Formula: AsFe2 | Molecular Weight: 186.612 g/mol

Iron arsenide is used as a superconductor, semiconductor and in photo optic applications 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12005-88-8
Molecular Formula AsFe2
Molecular Weight (g/mol) 186.612
MDL Number MFCD00151533
InChI Key FDKAYGUKROYPRO-UHFFFAOYSA-N
Synonym 1-arsa-2,3-diferracycloprop-1-ene, iron arsenide, pieces trace metals basis
PubChem CID 71310806
SMILES [Fe].[Fe]=[As]

Specifications

Physical Form Lump
Quantity 10 g
Odor Odorless
UN Number UN1557
Solubility Information It is insoluble in water.
Formula Weight 186.62
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Iron arsenide
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 234-474-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only