missing translation for 'onlineSavingsMsg'
Learn More

BlueMed IRIS Shoe Covers

Disposable shoe covers are made of a proprietary fabric that offers a balance between strength, anti-skid, fluid resistance, and wear resistance.

Supplier:  BlueMed SHS1408CB

 View more versions of this product

Catalog No. 18-009-121

Only null left
Explore available promotions

Description

Description

  • Barrier protection
  • Strong and durable
  • Anti-skid
  • Waterproof
  • Low linting, conforms to cleanrooms up to level ISO 5
  • Seamless sole and sides
  • Disposable
  • Color: Light Blue
  • Available in universal and XL Sizes
  • Made in Canada
Specifications

Specifications

Universal
Yes
Single-use
Elastic Ankle
Ultrasonically Welded
300/Cs.
16.93 in.
6.69 in.
60 gsm
Controlled Environments Shoe Covers
Polypropylene, Polyethylene
Light Blue
Non-sterile
Anti-skid
GMP or Research Facilities, Pharmaceutical Production or Labs
150/Dispenser Boxes, 2 Boxes/Cs.
43 cm
17 cm
Liquid
Class 100 (ISO 5)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions