Learn More
Sklar Instruments Iris Scissors, 4-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA.
Catalog No. 501109762
Encompass
Iris Scissors, 4-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. This product is straight with smooth, sharp/sharp tips and a length of 4-1/2 inches.
Catalog No. 501109762 Supplier Sklar Instruments Supplier No. 471145
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Iris Scissors, 4-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. This product is straight with smooth, sharp/sharp tips and a length of 4-1/2 inches.