Learn More
World Precision Iris Scissors, 11.5cm, Straight, 12-pack, Disposable
Catalog No. 50379958
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50379958 Supplier World Precision Supplier No. WP50370812
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.