missing translation for 'onlineSavingsMsg'
Learn More

Iridium(III) Chloride Hydrate, TCI America™

Catalog No. I06161G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06161G Supplier TCI America Supplier No. I06161G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Iridium (III) chloride hydrate, used in catalysis and inorganic chemistry research.

  • Iridium (III) chloride hydrate an anhydrous powder has a molecular weight of 298.57 with a melting Point 763°C
  • Stored in room temperature, in a cool dark place

Chemical Identifiers

CAS 14996-61-3
Molecular Formula Cl3Ir
Molecular Weight (g/mol) 298.57
MDL Number MFCD00149750
InChI Key DANYXEHCMQHDNX-UHFFFAOYSA-K
Synonym Iridium Trichloride Hydrate
IUPAC Name iridium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ir+3]

Specifications

Melting Point 763°C
Color Black-Yellow
Physical Form Crystalline Powder
Quantity 1 g
Linear Formula IrCl3·xH2O
Formula Weight 298.57
Concentration or Composition (by Analyte or Components) Iridium: 50.0% to 56.0%, Water: 15.0%
Manufacturing Origin Japan
Packaging Glass Bottle
Appearance Yellow to Dark Green to Black Powder to Crystal
Chemical Name or Material Iridium(III) Chloride Hydrate, Crystalline Powder
Show More Show Less