missing translation for 'onlineSavingsMsg'
Learn More

Iridium black, 99.95% (metals basis)

CAS: 7439-88-5 | Ir | 192.22 g/mol

$11188.51 - $11188.51

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4715009
View Documents
Thermo Scientific Chemicals
04715009
10 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-88-5
192.22
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
iridium
Ir
MFCD00011062
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
CHEBI:49666
[Ir]

Specifications

Black
10 g
MFCD00011062
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
[Ir]
192.22
CHEBI:49666
Iridium black
7439-88-5
Ir
UN3089
GKOZUEZYRPOHIO-UHFFFAOYSA-N
iridium
23924
99.95%
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q

H228-H319-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 231-095-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only