missing translation for 'onlineSavingsMsg'
Learn More

Irbesartan 98.0+%, TCI America™

Catalog No. I08595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08595G 5 g
I08591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08595G Supplier TCI America Supplier No. I08595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138402-11-6
Molecular Formula C25H28N6O
Molecular Weight (g/mol) 428.54
MDL Number MFCD00864464
InChI Key YOSHYTLCDANDAN-UHFFFAOYSA-N
PubChem CID 3749
ChEBI CHEBI:5959
IUPAC Name 2-butyl-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]-1,3-diazaspiro[4.4]non-1-en-4-one
SMILES CCCCC1=NC2(CCCC2)C(=O)N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NNN=N5

Specifications

Melting Point 188°C
Color White
Quantity 5 g
Formula Weight 428.54
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Irbesartan
RTECSNumber HM2950270
TSCA No