missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Irbesartan

Catalog No. AC465250010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465250010 1 g
AC465250050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465250010 Supplier Thermo Scientific Chemicals Supplier No. 465250010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 138402-11-6 | C25H28N6O | 428.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 138402-11-6
Molecular Formula C25H28N6O
Molecular Weight (g/mol) 428.54
InChI Key YOSHYTLCDANDAN-UHFFFAOYSA-N
IUPAC Name 2-butyl-3-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1,3-diazaspiro[4.4]non-1-en-4-one
SMILES CCCCC1=NC2(CCCC2)C(=O)N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1

Specifications

Melting Point 180.0°C to 182.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in dmso
Formula Weight 428.53
Chemical Name or Material Irbesartan

RUO – Research Use Only