missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IR-813 p-Toluenesulfonate 98.0+%, TCI America™
Supplier: TCI America C28861G
Specifications
| IR-813 p-Toluenesulfonate | |
| C47H47ClN2O3S | |
| 1 g | |
| LQVQHHTYXSPHSQ-UHFFFAOYSA-M | |
| 2-{2-[2-chloro-3-(2-{1,1,3-trimethyl-1H,2H,3H-benzo[e]indol-2-ylidene}ethylidene)cyclohex-1-en-1-yl]ethenyl}-1,1,3-trimethyl-1H-benzo[e]indol-3-ium 4-methylbenzene-1-sulfonate | |
| 91972145 | |
| ≥98.0% (HPLC,N) |
| 134127-48-3 | |
| MFCD03093252 | |
| 2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,1,3-trimethyl-2H-benz[e]indol-2-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-1,1,3-trimethyl-1H-benz[e]indolium 4-Methylbenzenesulfonate, 2-Chloro-3-[2-(1,1,3-trimethylbenz[e]indolin-2-ylidene)ethylidene]-1-[2-(1,1,3-t | |
| CC1=CC=C(C=C1)S([O-])(=O)=O.CN1C2=CC=C3C=CC=CC3=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=C4C=CC=CC4=C3C2(C)C)=C1Cl | |
| 755.41 | |
| 755.41 | |
| Crystalline Powder |
Chemical Identifiers
| 134127-48-3 | |
| 755.41 | |
| LQVQHHTYXSPHSQ-UHFFFAOYSA-M | |
| 91972145 | |
| CC1=CC=C(C=C1)S([O-])(=O)=O.CN1C2=CC=C3C=CC=CC3=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=C4C=CC=CC4=C3C2(C)C)=C1Cl |
| C47H47ClN2O3S | |
| MFCD03093252 | |
| 2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,1,3-trimethyl-2H-benz[e]indol-2-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-1,1,3-trimethyl-1H-benz[e]indolium 4-Methylbenzenesulfonate, 2-Chloro-3-[2-(1,1,3-trimethylbenz[e]indolin-2-ylidene)ethylidene]-1-[2-(1,1,3-t | |
| 2-{2-[2-chloro-3-(2-{1,1,3-trimethyl-1H,2H,3H-benzo[e]indol-2-ylidene}ethylidene)cyclohex-1-en-1-yl]ethenyl}-1,1,3-trimethyl-1H-benzo[e]indol-3-ium 4-methylbenzene-1-sulfonate |
Safety and Handling
EINECSNumber : (5)-6789
TSCA : No