missing translation for 'onlineSavingsMsg'
Learn More

IQGAP1 Recombinant Rabbit Monoclonal Antibody (23GB2020)

Catalog No. MA552473
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552473 20 μL
MA552472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552473 Supplier Fisher Chemical Supplier No. MA552473

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

IQGAP1 Recombinant Rabbit Monoclonal Antibody (23GB2020)

IQGAP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Specifications

Antigen IQGAP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2020
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IQGAP1
Gene Accession No. P46940, Q9JKF1
Gene Alias AA682088; Cdc42-Rac1 effector protein; D7Ertd237e; D7Ertd257e; DISP2; DISPB; HUMORFA01; IQ motif containing GTPase activating protein 1; IQGAP1; KIAA0051; mKIAA0051; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs; SAR1
Gene Symbols IQGAP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human IQGAP1 (1-58AA).
Purification Method Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29875, 361598, 8826
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less