missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ iQ Series™ Women's Lightweight Navy Flame Resistant Pant
iQ Series™ Women's Lightweight Navy Flame Resistant Pant is stylishly designed to keep you cool and in command all day long.
Supplier: Bulwark™ QP19NV1434U
Description
- Let our work pant do a little more of the lifting.
- The Bulwark™ iQ Series™ Lightweight Comfort Pant is stylishly designed to keep you cool and in command all day long.
- This modern, straight-fit jean style work pant features a one-piece waistband with wide belt loops, scoop front pockets, reinforced cargo pocket with snap closure on the left and patch hip pockets, a utility pocket on the right side plus triple needle out and inseam for extra durabilty.
- Certifications Level 1: NFPA 70E, NFPA 2112, ASTM F2302
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 6
- Primary Closure: Concealed Button
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Body: 6.5 oz. (220 g/m&superscript_2;) TenCate Evolv™ Flame Resistant
- Blend: Body: 50% Aramid / 25% Modacrylic / 25% Lyocell
- Pocket: Two scoop front pockets, Two patch hip pockets, One cargo pocket with snap closure on left leg and one utility pocket on right leg
- Features: Triple-needle felled main seams, Reinforced hip pockets and cargo pocket, Jean style cut with one-piece waistband and concealed button closure
- Product Collection: iQ Series Comfort
- Protection: ATPV Arc Rating: 8.4
- Country of Origin: Imported, Made in USA Fabric
- Gender: Female
- Brand: Bulwark™
Specifications
| 14 | |
| Aramid, Modacrylic, Lyocell | |
| Stitched | |
| Reusable | |
| 8.4 | |
| 34 in. |
| Women's | |
| Navy | |
| Concealed Button | |
| Category 2 | |
| NFPA 70E, NFPA 2112, ASTM F2302 |