missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ iQ Series™ Women's Lightweight Navy Flame Resistant Pant

iQ Series™ Women's Lightweight Navy Flame Resistant Pant is stylishly designed to keep you cool and in command all day long.

Supplier:  Bulwark™ QP19NV1834U

 View more versions of this product

Catalog No. 17-204-408

Only null left
Explore available promotions

Description

Description

  • Let our work pant do a little more of the lifting.
  • The Bulwark™ iQ Series™ Lightweight Comfort Pant is stylishly designed to keep you cool and in command all day long.
  • This modern, straight-fit jean style work pant features a one-piece waistband with wide belt loops, scoop front pockets, reinforced cargo pocket with snap closure on the left and patch hip pockets, a utility pocket on the right side plus triple needle out and inseam for extra durabilty.
  • Certifications Level 1: NFPA 70E, NFPA 2112, ASTM F2302
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 6
  • Primary Closure: Concealed Button
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Body: 6.5 oz. (220 g/m&superscript_2;) TenCate Evolv™ Flame Resistant
  • Blend: Body: 50% Aramid / 25% Modacrylic / 25% Lyocell
  • Pocket: Two scoop front pockets, Two patch hip pockets, One cargo pocket with snap closure on left leg and one utility pocket on right leg
  • Features: Triple-needle felled main seams, Reinforced hip pockets and cargo pocket, Jean style cut with one-piece waistband and concealed button closure
  • Product Collection: iQ Series Comfort
  • Protection: ATPV Arc Rating: 8.4
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Female
  • Brand: Bulwark™
Specifications

Specifications

18
Aramid, Modacrylic, Lyocell
Stitched
Concealed Button
Category 2
NFPA 70E, NFPA 2112, ASTM F2302
Women's
Navy
6
Reusable
8.4
34 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions