missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ iQ Series™ Women's Lightweight Navy Flame Resistant Pant

Catalog No. 17203955
Change view
Click to view available options
Size:
0
10
12
14
16
18
2
20
22
24
6
8
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17203955 0
17204040 2
17203983 -
17204320 6
17204380 8
17204399 10
17204269 12
17204465 14
17203966 16
17204408 18
17204319 20
17204477 22
17204462 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 17203955 Supplier Bulwark™ Supplier No. QP19NV 00 34U

iQ Series™ Women's Lightweight Navy Flame Resistant Pant is stylishly designed to keep you cool and in command all day long.

  • Let our work pant do a little more of the lifting.
  • The Bulwark™ iQ Series™ Lightweight Comfort Pant is stylishly designed to keep you cool and in command all day long.
  • This modern, straight-fit jean style work pant features a one-piece waistband with wide belt loops, scoop front pockets, reinforced cargo pocket with snap closure on the left and patch hip pockets, a utility pocket on the right side plus triple needle out and inseam for extra durabilty.
  • Certifications Level 1: NFPA 70E, NFPA 2112, ASTM F2302
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 6
  • Primary Closure: Concealed Button
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Body: 6.5 oz. (220 g/m&superscript_2;) TenCate Evolv™ Flame Resistant
  • Blend: Body: 50% Aramid / 25% Modacrylic / 25% Lyocell
  • Pocket: Two scoop front pockets, Two patch hip pockets, One cargo pocket with snap closure on left leg and one utility pocket on right leg
  • Features: Triple-needle felled main seams, Reinforced hip pockets and cargo pocket, Jean style cut with one-piece waistband and concealed button closure
  • Product Collection: iQ Series Comfort
  • Protection: ATPV Arc Rating: 8.4
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Female
  • Brand: Bulwark™

Specifications

Size 0
Gender Women's
Material Aramid, Modacrylic, Lyocell
Color Navy
Seam Type Stitched
No. of Pockets 6
Closure Type Concealed Button
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 8.4
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F2302
Size (English) Inseam 34 in.
Show More Show Less