missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ iQ Series™ Men's Lightweight Navy Flame Resistant Pant

Catalog No. 17204071
Change view
Click to view available options
Size:
28 x 37 in.
30 x 37 in.
34 x 37 in.
36 x 37 in.
38 x 37 in.
40 x 37 in.
42 x 37 in.
44 x 37 in.
46 x 37 in.
48 x 37 in.
50 x 37 in.
52 x 37 in.
54 x 37 in.
56 x 37 in.
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204071 28 x 37 in.
17204046 30 x 37 in.
17204452 34 x 37 in.
17204290 36 x 37 in.
17204458 38 x 37 in.
17204395 40 x 37 in.
17204447 42 x 37 in.
17204455 44 x 37 in.
17203953 46 x 37 in.
17204037 48 x 37 in.
17204038 50 x 37 in.
17204080 52 x 37 in.
17204125 54 x 37 in.
17204147 56 x 37 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 17204071 Supplier Bulwark™ Supplier No. QP18NV 28 37U

iQ Series™ Men's Lightweight Navy Flame Resistant Pant is stylishly designed to keep you cool and in command all day long.

  • Let our work pant do a little more of the lifting.
  • The Bulwark™ iQ Series™ Men's Lightweight Comfort Pant is stylishly designed to keep you cool and in command all day long.
  • This modern, straight-fit jean style work pant features a one-piece waistband with wide belt loops, scoop front pockets, reinforced cargo pocket with snap closure on the left and patch hip pockets, a utility pocket on the right side plus triple needle out and inseam for extra durability.
  • Certifications Level 1: ASTM F2302, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 6
  • Primary Closure: Concealed Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.5 oz. (220 g/m&superscript_2;) TenCate Evolv™ Flame Resistant
  • Blend: Body: 50% Aramid / 25% Modacrylic / 25% Lyocell
  • Pocket: Two scoop front pockets, Two patch hip pockets, One cargo pocket with snap closure on left leg and one utility pocket on right leg
  • Features: Triple-needle felled main seams, Reinforced hip pockets and cargo pocket, Jean style cut with one-piece waistband and concealed button closure
  • Product Collection: iQ Series Comfort
  • Protection: ATPV Arc Rating: 8.4
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Male
  • Brand: Bulwark™

Specifications

Size 28 x 37 in.
Gender Men's
Material Aramid, Modacrylic, Lyocell
Color Navy
Seam Type Stitched
No. of Pockets 6
Closure Type Concealed Button
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 8.4
Certifications/Compliance ASTM F2302, NFPA 2112, NFPA 70E
Show More Show Less