missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ iQ Series™ Men's Comfort Knit Light Blue Flame Resistant Henley

Catalog No. p-200000049
Change view
Click to view available options
Size:
2X-Large (Long)
3X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Catalog No. Size
17203965 Small
17204304 Medium
17204301 Large
17203979 X-Large
17204280 2X-Large
17204176 3X-Large
17204377 4X-Large
17204190 Large (Long)
17204311 X-Large (Long)
17203970 2X-Large (Long)
17204093 3X-Large (Long)
11 options

Catalog No. 17203965

Supplier: Bulwark™ QT20BL RG S

Only null left

iQ Series™ Men's Comfort Knit Light Blue Flame Resistant Henley is CAT 2 AR/FR, NFPA 2112 compliant and a casual, comfortable and stylish part of your FR system. With raglan sleeves and underarm gussets for added mobility—so you can bend, reach, stretch and work better.

  • This CAT 2 AR/FR, NFPA™ 2112 compliant, lightweight henley is a casual, comfortable and stylish part of your FR system.
  • With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better.
  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Button
  • Wash Care: Home Wash
  • Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m&superscript_2;)
  • Blend: 69% Cotton/ 25% Polyester/ 6% Polyoxadiazole
  • Closure: Four-button placket
  • Pocket: One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
  • Features: Moisture-wicking, breathable fabric, Raglan sleeves with full underarm gussets providing extra mobility, Hidden rib knit cuffs to keep sleeves in place
  • Product Collection: iQ Series Comfort, iQ Series
  • Packaging: Polybag

Specifications

Size Small
Gender Men's
Material Cotton, Polyester, POD
Color Light Blue
Neck Style Henley
Sleeve Style Raglan Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less