missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ iQ Series™ Men's Comfort Knit Light Blue Flame Resistant Henley

Catalog No. 17203965
Change view
Click to view available options
Size:
2X-Large (Long)
3X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17203965 Small
17204304 Medium
17204301 Large
17203979 X-Large
17204280 2X-Large
17204176 3X-Large
17204377 4X-Large
17204190 Large (Long)
17204311 X-Large (Long)
17203970 2X-Large (Long)
17204093 3X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 17203965 Supplier Bulwark™ Supplier No. QT20BL RG S

iQ Series™ Men's Comfort Knit Light Blue Flame Resistant Henley is CAT 2 AR/FR, NFPA 2112 compliant and a casual, comfortable and stylish part of your FR system. With raglan sleeves and underarm gussets for added mobility—so you can bend, reach, stretch and work better.

  • This CAT 2 AR/FR, NFPA™ 2112 compliant, lightweight henley is a casual, comfortable and stylish part of your FR system.
  • With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better.
  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Button
  • Wash Care: Home Wash
  • Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m&superscript_2;)
  • Blend: 69% Cotton/ 25% Polyester/ 6% Polyoxadiazole
  • Closure: Four-button placket
  • Pocket: One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
  • Features: Moisture-wicking, breathable fabric, Raglan sleeves with full underarm gussets providing extra mobility, Hidden rib knit cuffs to keep sleeves in place
  • Product Collection: iQ Series Comfort, iQ Series
  • Packaging: Polybag

Specifications

Size Small
Gender Men's
Material Cotton, Polyester, POD
Color Light Blue
Neck Style Henley
Sleeve Style Raglan Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less