missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ iQ Series™ Men's Comfort Knit Khaki Flame Resistant Henley
iQ Series™ Men's Comfort Knit Khaki Flame Resistant Henley is CAT 2 AR/FR, NFPA™ 2112 compliant and a casual, comfortable and stylish part of your FR system.
$203.50 - $203.50
Specifications
| Gender | Men's |
|---|---|
| Material | Cotton, Polyester, POD |
| Color | Khaki |
| Neck Style | Henley |
| Sleeve Style | Raglan Sleeve |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-204-111
|
Bulwark™
QT20KHRGS |
Small |
Each for $203.50
|
|
|||||
|
17-204-425
|
Bulwark™
QT20KHRGM |
Medium |
Each for $203.50
|
|
|||||
|
17-204-108
|
Bulwark™
QT20KHRGL |
Large |
Each for $203.50
|
|
|||||
|
17-204-204
|
Bulwark™
QT20KHRGXL |
X-Large |
Each for $203.50
|
|
|||||
|
17-204-386
|
Bulwark™
QT20KHRGXXL |
2X-Large |
Each for $203.50
|
|
|||||
|
17-204-340
|
Bulwark™
QT20KHRG3XL |
3X-Large |
Each for $203.50
|
|
|||||
|
17-204-434
|
Bulwark™
QT20KHRG4XL |
4X-Large |
Each for $203.50
|
|
|||||
|
17-204-180
|
Bulwark™
QT20KHLNL |
Large (Long) |
Each for $203.50
|
|
|||||
|
17-204-437
|
Bulwark™
QT20KHLNXL |
X-Large (Long) |
Each for $203.50
|
|
|||||
|
17-204-097
|
Bulwark™
QT20KHLNXXL |
2X-Large (Long) |
Each for $203.50
|
|
|||||
|
17-204-087
|
Bulwark™
QT20KHLN3XL |
3X-Large (Long) |
Each for $203.50
|
|
|||||
Description
- This CAT 2 AR/FR, NFPA™ 2112 compliant, lightweight henley is a casual, comfortable and stylish part of your FR system.
- With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better.
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Button
- Wash Care: Home Wash
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m&superscript_2;)
- Blend: 69% Cotton/ 25% Polyester/ 6% Polyoxadiazole
- Closure: Four-button placket
- Pocket: One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
- Features: Moisture-wicking, breathable fabric, Raglan sleeves with full underarm gussets providing extra mobility, Hidden rib knit cuffs to keep sleeves in place
- Product Collection: iQ Series Comfort, iQ Series
- Packaging: Polybag
Specifications
| Men's | |
| Khaki | |
| Raglan Sleeve | |
| Button | |
| 2 | |
| ASTM F1506, NFPA 2112, NFPA 70E |
| Cotton, Polyester, POD | |
| Henley | |
| Knit | |
| Reusable | |
| Category 2 |