missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ iQ Series™ Men's Comfort Knit Dark Blue Flame Resistant Henley

Catalog No. 17204439
Change view
Click to view available options
Size:
2X-Large (Long)
3X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204439 3X-Large (Long)
17204339 Small
17204430 Medium
17204113 Large
17204082 X-Large
17204420 2X-Large
17204383 3X-Large
17204179 4X-Large
17203992 Large (Long)
17204384 X-Large (Long)
17103754 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 17204439 Supplier Bulwark™ Supplier No. QT20DB LN 3XL

iQ Series™ Men's Comfort Knit Dark Blue Flame Resistant Henley is CAT 2 AR/FR, NFPA™ 2112 compliant and a casual, comfortable and stylish part of your FR system.

  • With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better
  • Four-button placket
  • One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
  • Moisture-wicking, breathable fabric
  • Hidden rib knit cuffs to keep sleeves in place
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Home Wash
  • Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m2)
  • Blend: 69% Cotton/25% Polyester/6% Polyoxadiazole
  • Packaging: Polybag

Specifications

Size 3X-Large (Long)
Gender Men's
Material Cotton, Polyester, POD
Color Navy
Neck Style Henley
Sleeve Style Raglan Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Fire
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less