missing translation for 'onlineSavingsMsg'
Learn More

IPTG, Dioxane-Free, Promega™

Catalog No. PRV3955
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3955 1 g
PRV3951 5 g
PRV3953 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRV3955 Supplier Promega Corporation Supplier No. V3955

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An inducer of beta-galactosidase activity in many bacteria.

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, unii-x73vv2246b, iptg dioxane free
PubChem CID 656894
ChEBI CHEBI:61448
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Quantity 1 g
Formula Weight 238.31
Percent Purity 99%
Grade Not Available
Physical Form Powder
Chemical Name or Material IPTG, Dioxane-Free, Moisture: ≤1% Dioxane Content: ≤10ppm
Recommended Storage Store dry at 4°C or -20°C.