missing translation for 'onlineSavingsMsg'
Learn More

IPTG, Dioxane-Free, Promega™

An inducer of beta-galactosidase activity in many bacteria.

Supplier:  Promega Corporation V3951

Encompass
Quantity 5 g

 View more versions of this product

Catalog No. PRV3951


Only null left

Description

Description

Specifications

Chemical Identifiers

367-93-1
238.30
BPHPUYQFMNQIOC-NXRLNHOXSA-N
656894
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
C9H18O5S
MFCD00063273
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, unii-x73vv2246b, iptg dioxane free
CHEBI:61448
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

IPTG, Dioxane-Free
367-93-1
C9H18O5S
5 g
BPHPUYQFMNQIOC-NXRLNHOXSA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
656894
238.31
Moisture: ≤1% Dioxane Content: ≤10ppm
White
MFCD00063273
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, unii-x73vv2246b, iptg dioxane free
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
238.30
CHEBI:61448
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store dry at 4°C or -20°C.

SDS
Documents

Documents

Promotions

Promotions