missing translation for 'onlineSavingsMsg'
Learn More

Ipratropium Bromide Monohydrate 98.0+%, TCI America™

Catalog No. p-7131649
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0907200MG 200 mg
I09071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0907200MG Supplier TCI America Supplier No. I0907200MG
Only null left

Chemical Identifiers

CAS 66985-17-9
Molecular Formula C20H32BrNO4
Molecular Weight (g/mol) 430.383
MDL Number MFCD09842549
InChI Key KEWHKYJURDBRMN-IUZMIPGZSA-M
Synonym Atropine Isopropyl Bromide
PubChem CID 129318280
IUPAC Name [(1R,5S)-8-methyl-8-propan-2-yl-8-azoniabicyclo[3.2.1]octan-3-yl] (2R)-3-hydroxy-2-phenylpropanoate;bromide;hydrate
SMILES CC(C)[N+]1(C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3)C.O.[Br-]

Specifications

Melting Point 231°C
Color White
Quantity 200 mg
Formula Weight 412.37
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ipratropium Bromide Monohydrate
RTECSNumber YM3700000
TSCA No