Learn More
Ipratropium bromide, 98%
CAS: 22254-24-6 | C20H30BrNO3 | 412.37 g/mol
Supplier: Thermo Scientific Chemicals J60453MC
Description
Muscarinic acetylcholine receptor antagonist and a bronchodilator
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ipratropium bromide | |
| 229°C to 231°C | |
| C20H30BrNO3 | |
| 100 mg | |
| ipratropium bromide hydrate, ipratropium bromide monohydrate, sch 1000-br-monohydrate, atrovent tn, atropine isopropyl bromide, unii-j697uz2a9j, 1r,3s,5s,8r-3-3-hydroxy-2-phenylpropanoyl oxy-8-isopropyl-8-methyl-8-azabicyclo 3.2.1 octan-8-ium hydrate bromide, ipratropium bromide usan:usp:inn:ban:jan, ipratropium bromide usan, sch-1000-br monohydrate | |
| [Br-].CC(C)[N+]1(C)[C@H]2CC[C@@H]1CC(C2)OC(=O)C(CO)C1=CC=CC=C1 | |
| 412.37 | |
| 412.37 | |
| Powder |
| 22254-24-6 | |
| White | |
| MFCD00069291 | |
| 14,5073 | |
| LHLMOSXCXGLMMN-CLTUNHJMSA-M | |
| (1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-methyl-8-(propan-2-yl)-8-azabicyclo[3.2.1]octan-8-ium bromide | |
| 16738693 | |
| 98% |
Chemical Identifiers
| 22254-24-6 | |
| 412.37 | |
| LHLMOSXCXGLMMN-CLTUNHJMSA-M | |
| 16738693 | |
| [Br-].CC(C)[N+]1(C)[C@H]2CC[C@@H]1CC(C2)OC(=O)C(CO)C1=CC=CC=C1 |
| C20H30BrNO3 | |
| MFCD00069291 | |
| ipratropium bromide hydrate, ipratropium bromide monohydrate, sch 1000-br-monohydrate, atrovent tn, atropine isopropyl bromide, unii-j697uz2a9j, 1r,3s,5s,8r-3-3-hydroxy-2-phenylpropanoyl oxy-8-isopropyl-8-methyl-8-azabicyclo 3.2.1 octan-8-ium hydrate bromide, ipratropium bromide usan:usp:inn:ban:jan, ipratropium bromide usan, sch-1000-br monohydrate | |
| (1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-methyl-8-(propan-2-yl)-8-azabicyclo[3.2.1]octan-8-ium bromide |
Safety and Handling
GHS H Statement
H302-H332-H319
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302+H332-H319
EINECSNumber : 244-873-8
RTECSNumber : YM3700000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only