Learn More
MilliporeSigma IPAK Quanta™ Polishing Cartridge for Purified Water
Catalog No. IPAKQUA00
Change view
Click to view available options
Feedwater Source:
Purified Water
Catalog No. Feedwater Source
IPAKQUA00 Purified Water
1 options
Catalog No. IPAKQUA00 Supplier MilliporeSigma Supplier No. IPAKQUA00
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For Milli-Q™ IQ/EQ 7000 and EQ 7008 7016 ultrapure water systems fed with purified water. This product replaces IPAKQUAA1.
  • e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with a drain cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal.
  • 78% of the parts suppliers are ISO 14001 environmental management system-certified.
Specifications
Feedwater Source Purified Water
Purification Method Ultrapure
Resin Type Ion Exchange Resin
Type Polishing Cartridge