Learn More
MilliporeSigma IPAK Quanta™ Polishing Cartridge for Elix™ Purified Water
Catalog No. IPAKQUAEX
Change view
Click to view available options
Feedwater Source:
Elix Purified Water
Catalog No. Feedwater Source
IPAKQUAEX Elix Purified Water
1 options
Catalog No. IPAKQUAEX Supplier MilliporeSigma Supplier No. IPAKQUAEX
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For Milli-Q™ IQ/EQ 7000 and IQ 7003/05 /10 /15 ultrapure water systems fed by Elix™ pure water. This product replaces IPAKQUAA1.
  • e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with a drain cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal.
  • 78% of the parts suppliers are ISO 14001 environmental management system certified.
Specifications
Feedwater Source Elix Purified Water
Purification Method Ultrapure
Resin Type Ion Exchange Resin
Type Polishing Cartridge