missing translation for 'onlineSavingsMsg'
Learn More

iota-Carrageenan, TCI America™

Catalog No. c180525g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C180525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C180525G Supplier TCI America Supplier No. C180525G

Chemical Identifiers

CAS 9062-07-1
Molecular Formula (C12H16O15S2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00151512
InChI Key QIDSWKFAPCTSKL-RRQHLKGPSA-J
Synonym iota-carrageenans, 1->4-3,6-anhydro-2-o-sulfonato-alpha-d-galactopyranosyl-1->3-4-o-sulfonato-beta-d-galactopyranan
PubChem CID 11966245
IUPAC Name (1R,3R,4R,5S,8S)-8-{[(2S,3R,4R,5R,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-(sulfonatooxy)oxan-2-yl]oxy}-3-{[(2S,3R,4R,5S,6R)-3-hydroxy-2-{[(1R,3S,4R,5S,8S)-3-hydroxy-4-(sulfonatooxy)-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)-5-(sulfonatooxy)oxan-4-yl]oxy}-2,6-dioxabicyclo[3.2.1]octan-4-yl sulfate
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H]3CO[C@@H]2[C@@H](OS(O)(=O)=O)[C@@H](-*)O3)[C@H](O)[C@@H](O-*)[C@H]1OS(O)(=O)=O

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material iota-Carrageenan
RTECSNumber FI0700000
TSCA No