Learn More
beta-Ionone, 96%, synthetic
CAS: 14901-07-6 | C13H20O | 192.30 g/mol
Supplier: Thermo Scientific Chemicals 297131000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| β-Ionone | |
| 14901-07-6 | |
| 100.0 | |
| 0.9440g/mL | |
| 122°C | |
| 95% min. (GC) | |
| C13H20O | |
| MFCD00001549 | |
| 07, 167 | |
| 10, 4914 | |
| Solubility in water: slightly soluble. Other solubilities: solutle in 2 to 3 parts of 70% alcohol, ether,, chloroform and benzene, miscible with absolute alcohol | |
| CC(=O)\C=C\C1=C(C)CCCC1(C)C | |
| 192.30 | |
| CHEBI:32325 | |
| 96% |
| 96% | |
| 95.0 | |
| Yellow | |
| 237.0°C to 239.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5190 to 1.5215 | |
| 100 g | |
| 0.944 | |
| beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone | |
| PSQYTAPXSHCGMF-BQYQJAHWSA-N | |
| (E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one | |
| 638014 | |
| 192.3 | |
| Liquid |
Chemical Identifiers
| 14901-07-6 | |
| 192.30 | |
| PSQYTAPXSHCGMF-BQYQJAHWSA-N | |
| 638014 | |
| (E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one |
| C13H20O | |
| MFCD00001549 | |
| beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone | |
| CHEBI:32325 | |
| CC(=O)\C=C\C1=C(C)CCCC1(C)C |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber : 238-969-9
RTECSNumber : EN0350000
TSCA : TSCA
RUO – Research Use Only