Description
The Ion PI™ Chip Kit v3 contains 8 barcoded chips for sample tracking and sequencing with the Ion Proton™ System. The Ion PI™ Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this revolutionary advance in next-generation sequencing technology allows for rapid sequencing times of less than 4 hours. The Ion PI™ Chip v3 enables a simplified sequencing workflow with the removal of chip preparation steps, reducing the hands-on time by approximately 45 minutes compared to previous versions.
The Ion PI™ Chip Kit v3 offers:
• A simplified workflow, reducing hands-on time by approximately 45 minutes
• Barcoded sequencing chip with sample tracking functionality
• 10 Gb of sequence data with a rapid run time of typically only 2-4 hours
• Support for a variety of applications, including sequencing of up to three human exomes, transcriptome profiling with 60-80 million reads, and targeted resequencing of gene panels
Notes
This kit is only compatible with the Ion Proton™ System.
Use this kit with the Ion PI™ Hi-Q OT2 200 Kit and Ion PI™ Hi-Q Sequencing 200 Kit.
This kit replaces the Ion PI™ Chip Kit v2 & Ion PI™ Chip Kit v2 BC.
Chemical Identifiers
| Ion PI | |
| Ion Proton™ System | |
| Chip Kit V3 |
| Store at room temperature. | |
| 60-80M | |
| Room Temperature |
Specifications
| Ion PI | |
| Ion Proton™ System | |
| Chip Kit V3 |
| Store at room temperature. | |
| 60-80M | |
| Room Temperature |