missing translation for 'onlineSavingsMsg'
Learn More

Ion exchanger I, H+ form, Reag. Ph Eur, EMD-Millipore

Catalog No. M1047650500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1047650500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1047650500 Supplier Technidata Supplier No. 1.04765.0500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Strongly acidic cation exchanger

Specifications

Chemical Name or Material Ion exchanger I
Density 850kg/m3
Quantity 500 g
Grade Reagent Ph. Eur.