Learn More
Solstice Ion Exchange SPE Columns, Strong Anion Exchange (Quaternary Amine Bonded Polymer), 50 mg/3 mL
Catalog No. 6003068
Change view
Click to view available options
Quantity:
50
Catalog No. Quantity
6003068 50
1 options
Catalog No. 6003068 Supplier Solstice Supplier No. 9088
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Medical-grade polypropylene with PE frits
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
Specifications
Quantity 50