Learn More
Ion Torrent™ Ion 550™ Single Chip Supplemental Kit
Description
The Ion 550 Single Chip Supplemental Kit contains pre-packaged templating supplies and sequencing reagent cartridges required for use with the single-chip Ion Chef workflow. The Ion 550 Single Chip Supplemental Kit, together with the Ion 550 Kit—Chef, enables the flexibility to prepare one loaded chip per Ion Chef run, without loss of templating reagents.
Note: This supplemental templating kit for Ion Chef is only compatible with the Ion 550 Chip and the Ion 550 Kit—Chef.
Related Links:
Specifications
Specifications
| For Use With (Application) | Comparative Sequencing (Germline Mutations - 50:50) |
| Quantity | 4 reactions |
| Product Line | Ion Torrent |
| For Use With (Equipment) | Ion 550 Chip, Ion 550 Kit-Chef |
| Product Type | Single Chip Supplemental Kit |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Content And Storage | 4 Ion S5 Chef Supplies, store at room temperature 1 Ion S5 Sequencing Solutions, store at room temperature 1 Ion S5 Sequencing Reagents, store at -20°C |
Frequently Asked Questions (FAQs)
Yes, Ion S5 Sequencing Reagents (Part No. A27768) is used on the instrument for both Ion 540 and Ion 550 Chip sequencing.
The Ion 550 Kit-Chef contains Ion Chef templating and sequencing consumables sufficient for preparing and sequencing eight Ion 550 chips for four Ion Chef runs and four sequencer initializations (dual chip run configuration). If you are running in the single chip configuration, then you will need to purchase the Ion 550 Single Chip Supplemental Kit separately to ensure that you have enough templating and sequencing consumables to support eight single chip Ion Chef templating runs and eight sequencer initializations with the Ion 550 Kit-Chef.
The Ion 550 Single Chip Supplemental Kit contains pre-packaged templating supplies and sequencing reagent cartridges required to support eight single chip Ion Chef runs and eight sequencer initializations with the Ion 550 Kit-Chef. The Ion 550 Single Chip Supplemental Kit includes the following components:
- Ion S5 Chef Supplies (4 boxes) (ships at room temperature; store at room temperature)
- Ion S5 Sequencing Solutions (1 box) (ships at room temperature; store at room temperature)
- Ion S5 Sequencing Reagents (1box) (ships on wet ice at 2-8 degrees C; store at -20 degrees C)
IMPORTANT! Do not store the Ion S5 Sequencing Reagents (Part No. A27768) on dry ice or in a closed environment containing dry ice.