missing translation for 'onlineSavingsMsg'
Learn More

Iohexol (mixture of isomers) 98.0+%, TCI America™

Catalog No. p-7131646
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09035G 5 g
I090325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09035G Supplier TCI America Supplier No. I09035G

Chemical Identifiers

CAS 66108-95-0
Molecular Formula C19H26I3N3O9
Molecular Weight (g/mol) 821.142
MDL Number MFCD00077732
InChI Key NTHXOOBQLCIOLC-UHFFFAOYSA-N
PubChem CID 3730
ChEBI CHEBI:31709
IUPAC Name 5-[acetyl(2,3-dihydroxypropyl)amino]-1-N,3-N-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide
SMILES CC(=O)N(CC(CO)O)C1=C(C(=C(C(=C1I)C(=O)NCC(CO)O)I)C(=O)NCC(CO)O)I

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Synonym N,N′C-Bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodoisophthalamide
Formula Weight 821.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Iohexol (mixture of isomers)
RTECSNumber CZ2205000
TSCA No