missing translation for 'onlineSavingsMsg'
Learn More

Iodomethylcyclopentane, 98%, stabilized

Catalog No. p-4258682
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378640010 1 g Glass bottle
AC378640050 5 g Glass bottle
AC378640250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC378640010 Supplier Thermo Scientific Chemicals Supplier No. 378640010

CAS: 27935-87-1 | C6H11I | 210.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27935-87-1
Molecular Formula C6H11I
Molecular Weight (g/mol) 210.06
MDL Number MFCD03453190
InChI Key DUMSKQUKLVSSII-UHFFFAOYSA-N
Synonym iodomethyl cyclopentane, cyclopentylmethyl iodide, cyclopentane, iodomethyl, cyclopentyliodomethane, acmc-20akpz, cyclopentylmethyliodide, iodomethyl-cyclopentane, cyclopentanemethyl iodide
PubChem CID 2762531
IUPAC Name iodomethylcyclopentane
SMILES ICC1CCCC1

Specifications

Boiling Point 78.0°C (17.0 mmHg)
Color Brown or Colorless to Orange
Physical Form Solution
Quantity 1 g
Assay Percent Range 97.5% min. (GC)
Linear Formula (C5H9)CH2I
Flash Point 92°C
Solubility Information Solubility in water: immiscible
Formula Weight 210.05
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5400 to 1.5440
Specific Gravity 1.6
Density 1.6000g/mL
Chemical Name or Material Iodomethylcyclopentane, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Recommended Storage May darken on storage

RUO – Research Use Only