missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)trimethylsilane 97.0+%, TCI America™

Catalog No. I044910G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I044910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I044910G Supplier TCI America Supplier No. I044910G

Chemical Identifiers

CAS 4206-67-1
Molecular Formula C4H11ISi
Molecular Weight (g/mol) 214.121
MDL Number MFCD00001077
InChI Key VZNYXGQMDSRJAL-UHFFFAOYSA-N
Synonym iodomethyl trimethylsilane, iodomethyl-trimethylsilane, iodomethyl trimethyl silane, trimethylsilyl methyl iodide, sila-neopentyliodide, trimethylsilylmethyl iodide, acmc-209jn7, trimethyl iodomethyl silane, iodanylmethyl trimethyl silane
PubChem CID 77877
IUPAC Name iodomethyl(trimethyl)silane
SMILES C[Si](C)(C)CI

Specifications

Color Yellow
Boiling Point 142°C
Quantity 10 g
UN Number 2924
Formula Weight 214.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (Iodomethyl)trimethylsilane