missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)trimethylammonium iodide, 98+%

Catalog No. p-7024161
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1842406 5 g
AAA1842414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1842406 Supplier Thermo Scientific Chemicals Supplier No. A1842406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39741-91-8 | C4H11I2N | 326.948 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39741-91-8
Molecular Formula C4H11I2N
Molecular Weight (g/mol) 326.948
MDL Number MFCD00031647
InChI Key IFVIWWQMNYZYFQ-UHFFFAOYSA-M
Synonym iodomethyltrimethylammonium iodide, iodomethyl trimethylammonium iodide, ammonium, iodomethyltrimethyl-, iodide, iodomethyl trimethylazanium iodide, 1-iodo-n,n,n-trimethylmethanaminium iodide, acmc-20akpy, iodomethyltrimethylammoniumiodide, iodomethyl trimethylamine, iodide, iodomethyl trimethyl azanium iodide, iodomethyl trimethylammonium iodide, 98+%
PubChem CID 38328
IUPAC Name iodomethyl(trimethyl)azanium;iodide
SMILES C[N+](C)(C)CI.[I-]

Specifications

Melting Point ∼190°C (decomposition)
Quantity 5 g
Beilstein 3912137
Sensitivity Hygroscopic; Light sensitive
Formula Weight 326.94
Percent Purity ≥98%
Chemical Name or Material (Iodomethyl)trimethylammonium iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 254-611-4
RTECSNumber BR6475000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only