missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)cyclopropane, 95%

Catalog No. p-7032892
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH35309MD 250 mg
AAH3530903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH35309MD Supplier Thermo Scientific Chemicals Supplier No. H35309MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33574-02-6 | C4H7I | 182.004 g/mol

(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 33574-02-6
Molecular Formula C4H7I
Molecular Weight (g/mol) 182.004
MDL Number MFCD06410734
InChI Key JHKJQWFHNIOUKY-UHFFFAOYSA-N
Synonym iodomethyl cyclopropane, cyclopropylmethyl iodide, cyclopropane, iodomethyl, ambkkkkk549, acmc-20aplj, 2-iodomethyl cyclopropane, iodomethyl cyclopropane gc
PubChem CID 2763048
IUPAC Name iodomethylcyclopropane
SMILES C1CC1CI

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 182
Percent Purity 97%
Chemical Name or Material (Iodomethyl)cyclopropane
Hazard Category H319-H334
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P280i-P285-P304+P341-P305+P351+P338-P342+P311-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only