Learn More
Iodomethane, 99%, stab. with copper
Description
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.
Solubility
Miscible with water.
Notes
Light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and oxygen.
Specifications
Specifications
| Melting Point | -67°C |
| Density | 2.28 |
| Boiling Point | 41°C to 43°C |
| Odor | Pungent |
| Refractive Index | 1.53 |
| Quantity | 50 g |
| UN Number | UN2644 |
| Beilstein | 969135 |
| Sensitivity | Light sensitive |
| Merck Index | 14,6087 |
| Show More |
Safety and Handling
| Hazard Category | H301+H331-H312-H315-H319-H335-H351 |
| Hazard Statement | GHS H Statement H301-H331-H351-H312-H315-H335 Toxic if swallowed. Toxic if inhaled. Suspected of causing cancer. Harmful in contact with skin. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL IODIDE |
| EINECSNumber | 200-819-5 |
| RTECSNumber | PA9450000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only