missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane, 99%, stab. with copper

Catalog No. AAA1055930
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1055930 250 g
AAA1055918 50 g
AAA105590B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1055930 Supplier Thermo Scientific Chemicals Supplier No. A1055930

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 74-88-4 | CH3I | 141.94 g/mol

Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.

Solubility
Miscible with water.

Notes
Light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and oxygen.

Chemical Identifiers

CAS 74-88-4
Molecular Formula CH3I
Molecular Weight (g/mol) 141.94
MDL Number MFCD00001073
InChI Key INQOMBQAUSQDDS-UHFFFAOYSA-N
Synonym methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek
PubChem CID 6328
ChEBI CHEBI:39282
IUPAC Name iodomethane
SMILES CI

Specifications

Melting Point -67°C
Density 2.28
Boiling Point 41°C to 43°C
Odor Pungent
Refractive Index 1.53
Quantity 250 g
UN Number UN2644
Beilstein 969135
Sensitivity Light sensitive
Merck Index 14,6087
Solubility Information Miscible with water.
Formula Weight 141.94
Percent Purity 99%
Chemical Name or Material Iodomethane, Stabilized with copper
Show More Show Less
Hazard Category H301+H331-H312-H315-H319-H335-H351
Hazard Statement GHS H Statement
H301-H331-H351-H312-H315-H335
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL IODIDE
EINECSNumber 200-819-5
RTECSNumber PA9450000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only