missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodomethane-13C Solution, NMR Reference Standard, NMR Tube Size: 5 mm 8 in., MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | NMR Tube Size: 5 mm 8 in. |
| Concentration | 1% in chloroform-d (“100%”, 99.96 atom% D) |
| Linear Formula | 13 CH3I |
| MDL Number | MFCD00001075 |
| Quantity | 1 Ea. |
| Formula Weight | 142.93 |
| Grade | NMR Reference Standard |