missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodomesitylene Diacetate 98.0+%, TCI America™
$184.54 - $184.54
Chemical Identifiers
| CAS | 33035-41-5 |
|---|---|
| Molecular Formula | C13H17IO4 |
| MDL Number | MFCD00674196 |
| Synonym | 2-(Diacetoxyiodo)mesitylene, 2,4,6-Trimethyl(diacetoxyiodo)benzene, 2,4,6-Trimethyliodobenzene Diacetate |
Chemical Identifiers
| 33035-41-5 | |
| C13H17IO4 | |
| MFCD00674196 | |
| 2-(Diacetoxyiodo)mesitylene, 2,4,6-Trimethyl(diacetoxyiodo)benzene, 2,4,6-Trimethyliodobenzene Diacetate |
Specifications
| 33035-41-5 | |
| White-Yellow | |
| MFCD00674196 | |
| 2-(Diacetoxyiodo)mesitylene, 2,4,6-Trimethyl(diacetoxyiodo)benzene, 2,4,6-Trimethyliodobenzene Diacetate | |
| ≥98.0% (T) | |
| Iodomesitylene Diacetate |
| 168°C | |
| C13H17IO4 | |
| 5 g | |
| 364.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator